Hexadecyltrimethylammonium perchlorate ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H0735-1G H0735-5G
TCIAH0735-1GEA 95.7 EUR
TCIAH0735-1G TCIAH0735-5G
Hexadecyltrimethylammonium perchlorate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium perchlorate
Formule: C₁₉H₄₂ClNO₄
MDL Nummer: MFCD00144895
CAS nummer: 6941-37-3
UN: 1479
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR