Bis(cyclopentadienyl)hafnium dichloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H0914-1G H0914-25G H0914-5G
TCIAH0914-1GEA 50.6 EUR
TCIAH0914-1G TCIAH0914-25G TCIAH0914-5G
Bis(cyclopentadienyl)hafnium dichloride ≥98.0% (by titrimetric analysis)
Bis(cyclopentadienyl)hafnium dichloride
Formule: C₁₀H₁₀Cl₂Hf
MDL Nummer: MFCD00001438
CAS nummer: 12116-66-4
EINECS: 235-177-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a7f5d05b9470dbb78c466238d804f50278eedcb&vid=7a7f5d05b9470dbb78c466238d804f50278eedcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR