3-(4-Hydroxy-3-methoxyphenyl)-1-propanol ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H1011-1G
TCIAH1011-1GEA 212 EUR
TCIAH1011-1G
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol ≥98.0% (by HPLC)
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol
Formule: C₁₀H₁₄O₃
MDL Nummer: MFCD00016571
CAS nummer: 2305-13-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 59.0 to 65.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR