2-Hydroxy-5-nitro-6-picoline ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H1160-1G H1160-5G
TCIAH1160-1GEA 35.2 EUR
TCIAH1160-1G TCIAH1160-5G
2-Hydroxy-5-nitro-6-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-nitro-6-picoline
Formule: C₆H₆N₂O₃
MDL Nummer: MFCD00092010
CAS nummer: 28489-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c69eaf9b0d062db4c5fb87914696242156b258d0&vid=c69eaf9b0d062db4c5fb87914696242156b258d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 233.0 to 238.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR