Zoutzuur 2 mol/L water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Hydrochloric acid solution, Muriatic acid

H1203-500ML
TCIAH1203-500MLEA 17.6 EUR
TCIAH1203-500ML
Zoutzuur 2 mol/L water
Zoutzuur
Formule: HCl
MW: 36.46 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III
Merck Index: 14,04780
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbd3f6058e9b93d5fcba6ad8a380092fc0fa89e4&vid=11d7e822f2863b4933e222fb0bcb0ff7f2fd95f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Concentration (Neutralization Titration) 1.9 to 2.1 mol/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR