3',4'-Dimethoxy-2'-hydroxyacetophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H1205-25G H1205-5G
TCIAH1205-25GEA 326 EUR
TCIAH1205-25G TCIAH1205-5G
3',4'-Dimethoxy-2'-hydroxyacetophenone ≥98.0% (by GC)
3',4'-Dimethoxy-2'-hydroxyacetophenone
Formule: C₁₀H₁₂O₄
MDL Nummer: MFCD00016684
CAS nummer: 5396-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=479940c62ab16e8d6670642a8b9e66b21aaa06e9&vid=479940c62ab16e8d6670642a8b9e66b21aaa06e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR