Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate

H1506-200MG H1506-1G
TCIAH1506-200MGEA 37.4 EUR
TCIAH1506-200MG TCIAH1506-1G
Hydrocortisone 17-Butyrate ≥98.0% (by HPLC)
Hydrocortisone 17-Butyrate
Formule: C₂₅H₃₆O₆
MW: 432.56 g/mol
MDL Nummer: MFCD00083364
CAS nummer: 13609-67-1
EINECS: 237-093-4
Merck Index: 14,04787
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=454b621f55405ca9d4ee87f13413fe1311664a86&vid=454b621f55405ca9d4ee87f13413fe1311664a86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 208.0 to 212.0 deg-C
Specific rotation [a]20/D +48.0 to +52.0 deg (C=0.1,CHCl3) (calcd.on dried substance)
Drying loss max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR