Hygromycin B ≥80.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H1509-200MG H1509-1G
TCIAH1509-200MGEA 47.3 EUR
TCIAH1509-200MG TCIAH1509-1G
Hygromycin B ≥80.0% (by HPLC)
Hygromycin B
Formule: C₂₀H₃₇N₃O₁₃
Smeltpunt: 180°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06795479
CAS nummer: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I
Merck Index: 14,04852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87e5e6b9444ace3deb62b303f8a59927327c55c5&vid=87e5e6b9444ace3deb62b303f8a59927327c55c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) (CAD) min. 80.0 area%
Specific rotation [a]20/D +18.0 to +23.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR