2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H1574-1G H1574-5G
TCIAH1574-1GEA 38.5 EUR
TCIAH1574-1G TCIAH1574-5G
2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)
2-Hydrazino-4-(trifluoromethyl)pyrimidine
Formule: C₅H₅F₃N₄
MDL Nummer: MFCD00115112
CAS nummer: 197305-97-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d264bf75eeaefe81b54deab81a721fb3fce39dd6&vid=64724cc630a90caca8e389ac3e509a49e8d5628e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 99.0 to 103.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR