(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulphonium Hexafluoroantimonate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H1620-25G H1620-5G
TCIAH1620-25GEA 387 EUR
TCIAH1620-25G TCIAH1620-5G
(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulphonium Hexafluoroantimonate ≥98.0% (by HPLC)
(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulphonium Hexafluoroantimonate
Formule: C₁₅H₁₇F₆OSSb
CAS nummer: 141651-31-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR