1H,1H-Heptafluoro-n-butyl methacrylate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2,2,3,3,4,4,4-Heptafluorobutyl Methacrylate, 1H,1h-heptafluorobutylmethacrylate, Methacrylic acid 2,2,3,3,4,4,4-Heptafluorobutyl ester

Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

H1674-5G H1674-25G
TCIAH1674-5GEA 52.8 EUR
TCIAH1674-5G TCIAH1674-25G
1H,1H-Heptafluoro-n-butyl methacrylate ≥98.0% (by GC) stabilised
1H,1H-Heptafluoro-n-butyl methacrylate
Formule: C₈H₇F₇O₂
MW: 268.13 g/mol
Vlampunt: 37°C
MDL Nummer: MFCD00039251
CAS nummer: 13695-31-3
EINECS: 237-217-7
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR