Potassium indole-3-acetate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I0023-1G I0023-25G
TCIAI0023-1GEA 19.8 EUR
TCIAI0023-1G TCIAI0023-25G
Potassium indole-3-acetate ≥98.0% (by HPLC, total nitrogen)
Potassium indole-3-acetate
Formule: C₁₀H₈KNO₂
MDL Nummer: MFCD00066213
CAS nummer: 2338-19-4
EINECS: 219-049-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e116536d30d56f2e3a3f061797f0d1a9b00625f7&vid=e116536d30d56f2e3a3f061797f0d1a9b00625f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR