3-(2-Hydroxyethyl)indole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I0030-1G I0030-25G I0030-5G
TCIAI0030-1GEA 27.5 EUR
TCIAI0030-1G TCIAI0030-25G TCIAI0030-5G
3-(2-Hydroxyethyl)indole ≥98.0% (by GC)
3-(2-Hydroxyethyl)indole
Formule: C₁₀H₁₁NO
Kookpunt: 179°C
Smeltpunt: 55…57 °C
MDL Nummer: MFCD00005659
CAS nummer: 526-55-6
EINECS: 208-393-2
Merck Index: 14,09798
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=484d1bec8ce996c34b037c195412f72f56af8264&vid=484d1bec8ce996c34b037c195412f72f56af8264&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 60.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR