Isopentylamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

I0082-25ML I0082-500ML I0082-100ML
TCIAI0082-25MLEA 18.7 EUR
TCIAI0082-25ML TCIAI0082-500ML TCIAI0082-100ML
Isopentylamine ≥98.0% (by GC, titration analysis)
Isopentylamine
Formule: C₅H₁₃N
Kookpunt: 97°C
Smeltpunt: –68 °C
Dichtheid: 0,75 g/cm³ (20 °C)
Vlampunt: -1°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008203
CAS nummer: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: 3,II
Merck Index: 14,05112
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a9477bf00f499ebc6d894aee427dcd49e04f98&vid=faae898fa059b780c09c81af8dcb70db4d0fee2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR