Benzyl isoamyl ether ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I0084-25ML
TCIAI0084-25MLEA 57.2 EUR
TCIAI0084-25ML
Benzyl isoamyl ether ≥98.0% (by GC)
Benzyl isoamyl ether
Formule: C₁₂H₁₈O
Kookpunt: 236°C
Vlampunt: 110°C
MDL Nummer: MFCD00048371
CAS nummer: 122-73-6
EINECS: 204-570-3

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR