1-Naphthyl isocyanate ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

I0125-25ML I0125-5ML
TCIAI0125-25MLEA 171 EUR
TCIAI0125-25ML TCIAI0125-5ML
1-Naphthyl isocyanate ≥99.0% (by GC)
1-Naphthyl isocyanate
Formule: C₁₁H₇NO
Kookpunt: 269°C
Smeltpunt: 36 °C
Dichtheid: 1,18 g/cm³ (20 °C)
Vlampunt: 135°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003881
CAS nummer: 86-84-0
EINECS: 201-703-7
UN: 2206
ADR: 6.1,III
Merck Index: 14,06410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57d9eced9400a1c713ae805594f17cb6c06abe84&vid=57d9eced9400a1c713ae805594f17cb6c06abe84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR