3-Iodotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0216-25G I0216-500G
TCIAI0216-25GEA 39.6 EUR
TCIAI0216-25G TCIAI0216-500G
3-Iodotoluene ≥98.0% (by GC)
3-Iodotoluene
Formule: C₇H₇I
Kookpunt: 213°C
Smeltpunt: -27°C
Dichtheid: 1,6835 g/cm³ (20 °C)
Vlampunt: 82°C
MDL Nummer: MFCD00001050
CAS nummer: 625-95-6
EINECS: 210-918-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=827bfda778bb95e4620ba4710f402c69a674ac0c&vid=8fc6cfcae7e24f898813daa857800e56497ccfef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR