Imidazole ≥99.0% (by GC, titration analysis) for fluorimetric analysis

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Glyoxaline, 1H-Imidazole, Glyoxalamine

I0290-25G
TCIAI0290-25GEA 137 EUR
TCIAI0290-25G
Imidazole ≥99.0% (by GC, titration analysis) for fluorimetric analysis
Imidazole
Formule: C₃H₄N₂
Kookpunt: 256°C
Smeltpunt: 89…90 °C
Dichtheid: 1,18 g/cm³ (20 °C)
Vlampunt: 135 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005183
CAS nummer: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8,III
Merck Index: 14,04912
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f36d8bd46f6669875e9b14c372ee978e29510608&vid=1e331395861d6012f1a3a428540e4c4c088d6beb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 88.0 to 91.0 deg-C
Intensity of fluorescence to pass test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR