Tetrabutylammonium bromide ≥99.0% (by titrimetric analysis) for ion pair chromatography

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0365-25G I0365-500G I0365-100G
TCIAI0365-25GEA 24.2 EUR
TCIAI0365-25G TCIAI0365-500G TCIAI0365-100G
Tetrabutylammonium bromide ≥99.0% (by titrimetric analysis) for ion pair chromatography
Tetrabutylammonium bromide
Formule: C₁₆H₃₆BrN
MW: 322.37 g/mol
Smeltpunt: 103°C
MDL Nummer: MFCD00011633
CAS nummer: 1643-19-2
EINECS: 216-699-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=687210e083cca81d851fd2d8ea8d8ed27134b482&vid=a2355fc23cb47a6641cfde8d06206edfab1d1a03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Absorbance (230nm) max. 1.00
Absorbance (254nm) max. 0.20
Absorbance (400nm) max. 0.10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR