Isoxazole ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

I0372-5G
TCIAI0372-5GEA 38.5 EUR
TCIAI0372-5G
Isoxazole ≥95.0% (by GC)
Isoxazole
Formule: C₃H₃NO
Kookpunt: 95°C
Dichtheid: 1,08 g/cm³ (20 °C)
Vlampunt: 12°C
MDL Nummer: MFCD00003149
CAS nummer: 288-14-2
EINECS: 206-018-7
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15b5599090a92fac5a99b0500634f844f19742ce&vid=15b5599090a92fac5a99b0500634f844f19742ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR