α,α,α-Trifluoro-2-iodotoluene ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with copper chip

I0383-25G
TCIAI0383-25GEA 71.5 EUR
TCIAI0383-25G
α,α,α-Trifluoro-2-iodotoluene ≥98.0% (by GC) stabilised
α,α,α-Trifluoro-2-iodotoluene
Formule: C₇H₄F₃I
Kookpunt: 198°C
Dichtheid: 1,896 g/cm³ (26 °C)
Vlampunt: 81°C
MDL Nummer: MFCD00001041
CAS nummer: 444-29-1
EINECS: 207-147-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=395644e6bcd9d4af05984f247da349d4580ba07d&vid=395644e6bcd9d4af05984f247da349d4580ba07d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR