Methyl (S)-(+)-2-isocyanato-3-tert-butoxypropionate ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0473-1G
TCIAI0473-1GEA 158 EUR
TCIAI0473-1G
Methyl (S)-(+)-2-isocyanato-3-tert-butoxypropionate ≥96.0% (by GC)
Methyl (S)-(+)-2-isocyanato-3-tert-butoxypropionate
Formule: C₉H₁₅NO₄
MDL Nummer: MFCD00191536
CAS nummer: 145080-94-0

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 96.0 %
Specific rotation [a]20/D +12.0 to +16.0 deg (neat)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR