Isopropyl methyl sulphide ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

I0483-25G
TCIAI0483-25GEA 158 EUR
TCIAI0483-25G
Isopropyl methyl sulphide ≥97.0% (by GC)
Isopropyl methyl sulphide
Formule: C₄H₁₀S
MW: 90.18 g/mol
Kookpunt: 85°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00015035
CAS nummer: 1551-21-9
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR