Methyl indole-5-carboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methyl 1H-indole-5-carboxylate

I0545-1G I0545-5G
TCIAI0545-1GEA 26.4 EUR
TCIAI0545-1G TCIAI0545-5G
Methyl indole-5-carboxylate ≥98.0% (by GC)
Methyl indole-5-carboxylate
Formule: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Nummer: MFCD00153023
CAS nummer: 1011-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5023bd16d6bf825682a9bac227ba73d2f071d5c9&vid=5023bd16d6bf825682a9bac227ba73d2f071d5c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR