Methyl-4-iodobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0593-25G I0593-5G
TCIAI0593-25GEA 48.4 EUR
TCIAI0593-25G TCIAI0593-5G
Methyl-4-iodobenzoate ≥98.0% (by GC)
Methyl-4-iodobenzoate
Formule: C₈H₇IO₂
Kookpunt: 276 °C (1013 hPa)
Smeltpunt: 112 °C
Dichtheid: 2,02 g/cm³ (10 °C)
MDL Nummer: MFCD00016353
CAS nummer: 619-44-3
EINECS: 210-597-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f86d14e9bc7cd58475aa16035c7e59b7b59863&vid=31f86d14e9bc7cd58475aa16035c7e59b7b59863&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR