Indapamide ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I0730-1G I0730-5G
TCIAI0730-1GEA 38.5 EUR
TCIAI0730-1G TCIAI0730-5G
Indapamide ≥97.0% (by HPLC, titration analysis)
Indapamide
Formule: C₁₆H₁₆ClN₃O₃S
MDL Nummer: MFCD00079375
CAS nummer: 26807-65-8
EINECS: 248-012-7
Merck Index: 14,04935

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 167.0 to 173.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR