Iron(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I0783-200MG I0783-1G
TCIAI0783-200MGEA 58.3 EUR
TCIAI0783-200MG TCIAI0783-1G
Iron(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation
Iron(II) phthalocyanine
Formule: C₃₂H₁₆FeN₈
MDL Nummer: MFCD00015953
CAS nummer: 132-16-1
EINECS: 205-047-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=459012f0e78e6d48ba44545f9065d085803797f4&vid=7977bb38686bd7f09fa1acca9bc19220cb60b9cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark purple powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR