2-Iodo-4-(trifluoromethyl)aniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0794-25G I0794-5G
TCIAI0794-25GEA 170 EUR
TCIAI0794-25G TCIAI0794-5G
2-Iodo-4-(trifluoromethyl)aniline ≥98.0% (by GC)
2-Iodo-4-(trifluoromethyl)aniline
Formule: C₇H₅F₃IN
MDL Nummer: MFCD02683097
CAS nummer: 163444-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd17f1ffef0be689f1dd6d1893b3b235e6a64736&vid=dd17f1ffef0be689f1dd6d1893b3b235e6a64736&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR