4-Iodophenol ≥98.0% (by GC) for biochemical research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0840-5G I0840-1G
TCIAI0840-5GEA 44 EUR
TCIAI0840-5G TCIAI0840-1G
4-Iodophenol ≥98.0% (by GC) for biochemical research
4-Iodophenol
Formule: C₆H₅IO
Kookpunt: 287 °C (1013 hPa)
Smeltpunt: 92…93 °C
Dichtheid: 2,13 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002327
CAS nummer: 540-38-5
EINECS: 208-745-5
ADR: 8,III
Merck Index: 14,05036
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b481c6b5d4fd61bc54d9c752441e08ac9a00a31&vid=9083e9bd60106986f62d013e0845eca2ba86e040&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C
Chemiluminescence test (Luminol/POD/H2O2) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR