2-Methylpropeen ∼15% in THF

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

I0911-100ML I0911-500ML
TCIAI0911-100MLEA 27.5 EUR
TCIAI0911-100ML TCIAI0911-500ML
2-Methylpropeen ∼15% in THF
2-Methylpropeen
Formule: C₄H₈
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008898
CAS nummer: 115-11-7
EINECS: 204-066-3
UN: 1993
ADR: 3,II
Merck Index: 14,05141
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae0ab1b2f0e0764a78f04c29bdd934e23307eecb&vid=ae0ab1b2f0e0764a78f04c29bdd934e23307eecb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 14.0 to 16.0 w/w%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR