4-Isothiocyanato-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

I0987-1G I0987-5G
TCIAI0987-1GEA 63.8 EUR
TCIAI0987-1G TCIAI0987-5G
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile
Formule: C₉H₃F₃N₂S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09800709
CAS nummer: 143782-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f51471fdf149e7612ef676294fc5ddcf014147e&vid=9f51471fdf149e7612ef676294fc5ddcf014147e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR