Dimethyl-2-oxoglutarate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

K0013-25ML K0013-5ML
TCIAK0013-25MLEA 400 EUR
TCIAK0013-25ML TCIAK0013-5ML
Dimethyl-2-oxoglutarate ≥95.0% (by GC)
Dimethyl-2-oxoglutarate
Formule: C₇H₁₀O₅
Kookpunt: 117°C
MDL Nummer: MFCD00048052
CAS nummer: 13192-04-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR