N-Lauroylsarcosine ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L0151-25G L0151-500G L0151-100G
TCIAL0151-25GEA 17.6 EUR
TCIAL0151-25G TCIAL0151-500G TCIAL0151-100G
N-Lauroylsarcosine ≥90.0% (by GC)
N-Lauroylsarcosine
Formule: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH
MDL Nummer: MFCD00021749
CAS nummer: 97-78-9
EINECS: 202-608-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7b428c500bc10787606e0dedb3d1759e91cea90&vid=b7b428c500bc10787606e0dedb3d1759e91cea90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR