Lithium acetate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L0191-25G L0191-500G
TCIAL0191-25GEA 42.9 EUR
TCIAL0191-25G TCIAL0191-500G
Lithium acetate ≥98.0% (by titrimetric analysis)
Lithium acetate
Formule: H₃CCOOLi
Smeltpunt: 283°C
Dichtheid: 1,26 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013057
CAS nummer: 546-89-4
EINECS: 208-914-3
Merck Index: 14,05521
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb6d3c6b579711c5bc32d2fe833ee694a47c0bdc&vid=768c1c54421b79491588c7d509c907f5c938447c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR