Lithium bis(nonafluorobutanesulphonyl)imide ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L0307-1G L0307-5G
TCIAL0307-1GEA 59.4 EUR
TCIAL0307-1G TCIAL0307-5G
Lithium bis(nonafluorobutanesulphonyl)imide ≥95.0% (by titrimetric analysis)
Lithium bis(nonafluorobutanesulphonyl)imide
Formule: C₈F₁₈LiNO₄S₂
MDL Nummer: MFCD06200837
CAS nummer: 119229-99-1
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b05d5674463092d1abf02619587b40f0e107fc5e&vid=b05d5674463092d1abf02619587b40f0e107fc5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR