Monosodium maleate trihydrate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Sodium Hydrogen Maleate, Sodium Bimaleate, Maleic Acid Monosodium Salt

M0016-25G
TCIAM0016-25GEA 74.8 EUR
TCIAM0016-25G
Monosodium maleate trihydrate ≥98.0% (by titrimetric analysis)
Monosodium maleate trihydrate
Formule: C₄H₄O₄
MW: 138.05 g/mol
MDL Nummer: MFCD00070246
CAS nummer: 3105-55-3
EINECS: 221-461-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 26.0 to 30.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR