D-(+)-Malic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid

M0021-25G M0021-5G
TCIAM0021-25GEA 126 EUR
TCIAM0021-25G TCIAM0021-5G
D-(+)-Malic acid ≥98.0% (by GC, titration analysis)
D-(+)-Malic acid
Formule: C₄H₆O₅
MW: 134.09 g/mol
Kookpunt: 322
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004245
CAS nummer: 636-61-3
EINECS: 211-262-2
Merck Index: 14,05707
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27f3df43216597a07a310ac9fce30582fefa6e2b&vid=27f3df43216597a07a310ac9fce30582fefa6e2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR