Malonic acid ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1,3-Propanedioic acid

M0028-25G M0028-500G
TCIAM0028-25GEA 17.6 EUR
TCIAM0028-25G TCIAM0028-500G
Malonic acid ≥99.0% (by titrimetric analysis)
Malonic acid
Formule: C₃H₄O₄
MW: 104.06 g/mol
Kookpunt: 387
Smeltpunt: 136
Vlampunt: 172
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002707
CAS nummer: 141-82-2
EINECS: 205-503-0
Merck Index: 14,05710
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=842b72e84a565ff87999f4c58e058eca02142935&vid=842b72e84a565ff87999f4c58e058eca02142935&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR