Poly(methyl methacrylate)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methacrylic Acid Methyl Ester Polymer, Methyl Methacrylate Polymer, PMMA

M0088-25G M0088-500G
TCIAM0088-25GEA 17.6 EUR
TCIAM0088-25G TCIAM0088-500G
Poly(methyl methacrylate)
Poly(methyl methacrylate)
Formule: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL Nummer: MFCD00134349
CAS nummer: 9011-14-7

Order Now

Specification Test Results

Appearance White powder to crystal
Solubility in THF very faint turbidity
Viscosity 2.0 to 4.0 mPa-s (0.5g/50mL THF, 20 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR