L-Methionine ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M0099-25G M0099-500G M0099-100G
TCIAM0099-25GEA 19.8 EUR
TCIAM0099-25G TCIAM0099-500G TCIAM0099-100G
L-Methionine ≥99.0% (by HPLC, titration analysis)
L-Methionine
Formule: C₅H₁₁NO₂S
Kookpunt: 278 °C (1013 hPa)
Smeltpunt: 136 °C
Dichtheid: 1,373 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063097
CAS nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 14,05975
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dceb2fecd6ee6e3787a46f5364a8a563f14b951&vid=2dceb2fecd6ee6e3787a46f5364a8a563f14b951&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D +23.7 to +24.7 deg (C=8, HCl (1+1)) (after drying)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR