Methylandrostenediol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M0146-1G M0146-25G
TCIAM0146-1GEA 34.1 EUR
TCIAM0146-1G TCIAM0146-25G
Methylandrostenediol ≥98.0% (by GC)
Methylandrostenediol
Formule: C₂₀H₃₂O₂
MDL Nummer: MFCD00021126
CAS nummer: 521-10-8
EINECS: 208-301-0
Merck Index: 14,05950

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 202.0 to 207.0 deg-C
Specific rotation [a]20/D -71.0 to -77.0 deg (C=1, EtOH)
Solubility in EtOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR