3-Methyl-2-benzothiazolinonehydrazone hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

M0160-25G M0160-5G
TCIAM0160-25GEA 114 EUR
TCIAM0160-25G TCIAM0160-5G
3-Methyl-2-benzothiazolinonehydrazone hydrochloride ≥98.0% (by HPLC, titration analysis)
3-Methyl-2-benzothiazolinonehydrazone hydrochloride
Formule: C₈H₁₀ClN₃S
MDL Nummer: MFCD00005816
CAS nummer: 4338-98-1
EINECS: 224-393-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 % (after drying)
Solubility in hot Water almost transparency
Drying loss max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR