Methylenedisalicylic acid (mixture of isomers) ≥90.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M0221-25G M0221-500G
TCIAM0221-25GEA 17.6 EUR
TCIAM0221-25G TCIAM0221-500G
Methylenedisalicylic acid (mixture of isomers) ≥90.0% (by titrimetric analysis)
Methylenedisalicylic acid (mixture of isomers)
Formule: C₁₅H₁₂O₆
MDL Nummer: MFCD00016506
CAS nummer: 27496-82-8
EINECS: 248-497-5
Merck Index: 14,06065

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR