5-Methyl-3-heptanon ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Amyl ethyl ketone, Ethyl Amyl ketone

M0335-25ML M0335-500ML
TCIAM0335-25MLEA 17.6 EUR
TCIAM0335-25ML TCIAM0335-500ML
5-Methyl-3-heptanon ≥95.0% (by GC)
5-Methyl-3-heptanon
Formule: C₈H₁₆O
MW: 128.21 g/mol
Kookpunt: 157°C
Smeltpunt: -56.7
Vlampunt: 59°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009340
CAS nummer: 541-85-5
EINECS: 208-793-7
UN: 2271
ADR: 3 III
Merck Index: 14,06084
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22e364fdcb88d138ad1c3aea621703f79f9515db&vid=a738e0eb97a2936de033f9e6db869a90f83c103c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR