Diethyl methylmalonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diethyl 2-methylmalonate

M0366-25ML M0366-500ML M0366-100ML
TCIAM0366-25MLEA 17.6 EUR
TCIAM0366-25ML TCIAM0366-500ML TCIAM0366-100ML
Diethyl methylmalonate ≥98.0% (by GC)
Diethyl methylmalonate
Formule: C₈H₁₄O₄
MW: 174.2 g/mol
Kookpunt: 199°C
Vlampunt: 82°C
MDL Nummer: MFCD00009162
CAS nummer: 609-08-5
EINECS: 210-175-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060c0e09dbeea5cb73d85b9621b289eac1362512&vid=060c0e09dbeea5cb73d85b9621b289eac1362512&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR