Methyl red ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

M0421-1G M0421-25G
TCIAM0421-1GEA 18.7 EUR
TCIAM0421-1G TCIAM0421-25G
Methyl red ≥98.0% (by HPLC)
Methyl red
Formule: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002425
CAS nummer: 493-52-7
EINECS: 207-776-1
Merck Index: 14,06119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6f65b0d2c9aab4635f969772d5a834b022c0492&vid=b6f65b0d2c9aab4635f969772d5a834b022c0492&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Absorbance min. 0.350 (5 mg/L, EtOH, pH 6.6, 428.0 to 432.0 nm)
Solubility in hot EtOH (95vol%) transparency
Transition interval (pH) (purplish red)4.2 to 6.2 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR