Methyltriethoxysilane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M0451-25ML M0451-500ML
TCIAM0451-25MLEA 17.6 EUR
TCIAM0451-25ML TCIAM0451-500ML
Methyltriethoxysilane ≥98.0% (by GC)
Methyltriethoxysilane
Formule: C₇H₁₈O₃Si
Kookpunt: 143°C
Smeltpunt: –50 °C
Dichtheid: 0,89 g/cm³ (20 °C)
Vlampunt: 23°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009066
CAS nummer: 2031-67-6
EINECS: 217-983-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c90e7d8c9d82c58793cc0e9f94794090d17d847&vid=2c90e7d8c9d82c58793cc0e9f94794090d17d847&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR