5-Methyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M0452-1G M0452-100MG
TCIAM0452-1GEA 882 EUR
TCIAM0452-1G TCIAM0452-100MG
5-Methyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
5-Methyl-DL-tryptophan
Formule: C₁₂H₁₄N₂O₂
MDL Nummer: MFCD00005652
CAS nummer: 951-55-3
EINECS: 213-453-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR