Hexyl methacrylate ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

M0503-25ML M0503-500ML
TCIAM0503-25MLEA 33 EUR
TCIAM0503-25ML TCIAM0503-500ML
Hexyl methacrylate ≥98.0% (by GC) stabilised
Hexyl methacrylate
Formule: C₁₀H₁₈O₂
Kookpunt: 88°C
Vlampunt: 80°C
MDL Nummer: MFCD00015283
CAS nummer: 142-09-6
EINECS: 205-521-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR