3-Methylthiopropylamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 3-Aminopropyl Methyl Sulphide, 3-Methylmercaptopropylamine

M0754-25ML M0754-5ML
TCIAM0754-25MLEA 292 EUR
TCIAM0754-25ML TCIAM0754-5ML
3-Methylthiopropylamine ≥98.0% (by GC, titration analysis)
3-Methylthiopropylamine
Formule: C₄H₁₁NS
MW: 105.20 g/mol
Kookpunt: 170°C
Vlampunt: 62°C
MDL Nummer: MFCD00041898
CAS nummer: 4104-45-4
EINECS: 223-875-2
UN: 2735
ADR: 8 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR